Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p19ArfRat anti-Hamster, Human, Mouse, Clone: 5.C3.1, Rockland™
SDP

Catalog No. 501991222
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1222 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-1222 Supplier Rockland Immunochemicals Supplier No. 200501891
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Directed against mouse p19Arf protein. No reactivity is observed against human or hamster homologues. The epitope was putatively mapped to amino acids 62-75 of mouse p19Arf. BLAST analysis indicates that no significant sequence homology exists for this sequence with p19Arf homologues from other sources. The homologue from rat only shows 11 of 14 identities (78% homology). No additional information is available for reactivity with p19Arf protein from other sources.

In humans, the multiple tumor suppressor 1 locus (MTS1) encodes two unrelated genes, p16INK4a and p19ARF. Although both act as cell proliferation inhibitors, their mechanisms of action are different. p19ARF works as part of a p53-dependent pathway to counter uncontrolled proliferation and oncogenic signals. Mice lacking the p19ARF gene rapidly develop a broad spectrum of tumors. This result indicates that p19ARF is an important tumor suppressor. In addition, p19ARF helps prevent transforming signals from various onocoproteins via the p53 regulatory loop. These studies and others display the importance of p19ARF in cell cycle control and tumor suppression.

Specifications

Antigen p19Arf
Applications ELISA, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 5.C3.1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene Cdkn2a
Gene Accession No. P42771, P51480, Q64364, Q8N726
Gene Alias Alternative reading frame; ARF; ARF-INK4a; CDK4I; Cdkn2a; Cyclin-dependent kinase 4 inhibitor A; Cyclin-dependent kinase inhibitor 2A; cyclin-dependent kinase inhibitor 2A (p16, inhibits CDK4); cyclin-dependent kinase inhibitor 2A, isoform 3; cyclin-dependent kinase inhibitor 2A, isoforms 1/2; cyclin-dependent kinase inhibitor protein; Ink4a/Arf; INK4a-ARF; mitochondrial smARF; MTS1; p16; p16(INK4a); p16-INK4; P16ink4a; p16-INK4a; p19; p19ARF; Pctr1; Tumor suppressor ARF
Gene Symbols Cdkn2a
Host Species Rat
Immunogen This protein G purified monoclonal antibody was produced by repeated immunizations with a synthetic peptide corresponding to an internal region near amino acid residues 54-75 of mouse p19Arf protein.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100764983, 1029, 12578
Target Species Hamster, Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.