Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ p21 ARC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518275
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518275 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518275 Supplier Invitrogen™ Supplier No. PA518275
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

This gene encodes one of seven subunits of the human Arp2/3 protein complex. The Arp2/3 protein complex has been conserved through evolution and is implicated in the control of actin polymerization in cells. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p21 ARC
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ARPC3
Gene Accession No. O15145, Q9JM76
Gene Alias 1110006A04Rik; 21kDa; AA408672; actin related protein 2/3 complex subunit 3; actin related protein 2/3 complex subunit 3, 21kDa; actin related protein 2/3 complex, subunit 3; actin related protein 2/3 complex, subunit 3, 21kDa; actin-related protein 2/3 complex subunit 3; AI788639; ARC21; Arp2/3 complex 21 kDa subunit; Arp2/3 complex subunit p21-Arc; ARP2/3 protein complex subunit p21; Arpc3; p21-Ar; p21Arc; p21-Arc
Gene Symbols ARPC3
Host Species Goat
Immunogen Synthetic peptide sequence (RQFMNKSLSGPGQ) corresponding to the C-terminus amino acids of ARPC3 (aa 166-178).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10094, 56378
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.