Learn More
Invitrogen™ p21 ARC Polyclonal Antibody

Description
Recommended positive controls: The peptide used to generate this antibody is available for purchase (GTX44635-PEP).. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | p21 ARC |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | ARPC3 |
| Gene Accession No. | O15145, Q9JM76 |
| Gene Alias | 1110006A04Rik; 21kDa; AA408672; actin related protein 2/3 complex subunit 3; actin related protein 2/3 complex subunit 3, 21kDa; actin related protein 2/3 complex, subunit 3; actin related protein 2/3 complex, subunit 3, 21kDa; actin-related protein 2/3 complex subunit 3; AI788639; ARC21; Arp2/3 complex 21 kDa subunit; Arp2/3 complex subunit p21-Arc; ARP2/3 protein complex subunit p21; Arpc3; p21-Ar; p21Arc; p21-Arc |
| Gene Symbols | ARPC3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.