Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p21/CIP1/CDKN1A Antibody (SR1331), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | p21/CIP1/CDKN1A |
---|---|
Clone | SR1331 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Description
p21/CIP1/CDKN1A Monoclonal antibody specifically detects p21/CIP1/CDKN1A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
p21/CIP1/CDKN1A | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CAP20cyclin-dependent kinase inhibitor 1, CDK-interacting protein 1, CDKN1melanoma differentiation associated protein 6, CIP1WAF1CDK-interaction protein 1, cyclin-dependent kinase inhibitor 1A (p21, Cip1), MDA6, MDA-6, Melanoma differentiation-associated protein 6, p21, p21CIP1, p21Cip1/Waf1, PIC1, SDI1DNA synthesis inhibitor, wild-type p53-activated fragment 1 | |
A synthesized peptide derived from human p21/CIP1/CDKN1A (Uniprot #: P38936) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1331 | |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, DNA Repair, Phospho Specific | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
1026 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title