Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p21 Rabbit anti-Mouse, Unconjugated, Polyclonal, Proteintech
SDP

Catalog No. 502238647 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-223-8647 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-223-8647 Supplier Proteintech Group Inc Supplier No. 282481AP150UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The p21 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse p21. This antibody recognizes mouse, rat antigen. The p21 antibody has been validated for the following applications: IF, WB, ELISA analysis.

Specifications

Antigen p21
Applications Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
Classification Polyclonal
Concentration 467 μg/mL
Conjugate Unconjugated
Host Species Rabbit
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 12575
Target Species Mouse, Rat
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.