Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p21/Waf1/Cip1 Rat anti-Human,Mouse, Unconjugated, Clone: HUGO-291, MilliporeSigma™

Catalog No. MABE18162MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE18162MI 25 μg
MABE18161MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABE18162MI Supplier MilliporeSigma Supplier No. MABE181625UG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

p21/Waf1/Cip1 Monoclonal antibody specifically detects p21/Waf1/Cip1 in Human,Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p21/Waf1/Cip1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone HUGO-291
Conjugate Unconjugated
Gene Accession No. NP_031695.1
Gene Alias Cyclin-dependent kinase inhibitor 1;CDK-interacting protein 1;Melanoma differentiation-associated protein
Gene Symbols Cdkn1a, Cip1, Waf1
Host Species Rat
Immunogen GST/His-tagged full-length murine recombinant p21 protein.
Molecular Weight of Antigen ∼26 kDa observed; 17.78 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 25 μg
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity Clone HUGO-291 is a rat monoclonal antibody that specifically detects p21 protein.
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.