Learn More
Description
Host Species: Mouse
Clone: HJ21; same as WA-1
Isotype: IgG1
Species Reactivity: Human and Rat. Others not known.
Epitope: Not determined
Immunogen: Human recombinant p21 protein
Molecular Weight: 21kDa
Positive Control: PC12, Ls174T or Raji cells
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Not suitable)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Not suitable)
- Flow Cytometry (Not Suitable)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | p21WAF1 Ab-6 |
| Applications | Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | HJ21; same as WA-1 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Human recombinant p21 protein |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.