Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p21WAF1 Ab-6 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS454PABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS454PABX 200 μL 1mg/mL
MS454P0 100 μL 200μg/mL
MS454P1 500 μL 200μg/mL
MS454P 1 mL 200μg/mL
MS454P1ABX 100 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS454PABX Supplier Epredia Supplier No. MS454PABX
Add to Cart
Add to Cart

Achieve accurate, reproducible results with the Epredia™ p21WAF1 Ab-6, Mouse Monoclonal Antibody.

p21WAF1/Cip1/Sdi1/Pic1 is a tumor suppressor protein. Expression of p21WAF1 is induced by wild type, but not mutant, p53 suppressor protein. The p21WAF1 protein binds to cyclin/CDK complexes and inhibits their kinase activity thereby stopping cell cycle progression. It also binds to PCNA (proliferating cell nuclear antigen) and blocks DNA replication but not the DNA repair process.

Host Species: Mouse

Clone: HJ21; same as WA-1

Isotype: IgG1

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Human recombinant p21 protein

Molecular Weight: 21kDa

Positive Control: PC12, Ls174T or Raji cells

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Not suitable)
  • Flow Cytometry (Not Suitable)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen p21WAF1 Ab-6
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone HJ21; same as WA-1
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human recombinant p21 protein
Quantity 200 μL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human, Rat
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.