Learn More
Description
Host Species: Mouse
Clone: DCS-60.2
Isotype: IgG2a
Species Reactivity: Human. Does not react with mouse and rat. Others not known
Epitope: Not determined
Immunogen: Full-length human recombinant p21 protein
Molecular Weight: 21kDa
Positive Control: Ls174T or Raji cells, Colon CA
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting (Not suitable)
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
- Flow Cytometry (Not suitable)
Specifications
Specifications
| Antigen | p21WAF1 Ab-3 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation |
| Classification | Monoclonal |
| Clone | DCS-60.2 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Full length human recombinant p21 protein |
| Quantity | 200 μL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.