Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p23/PTGES3 Antibody (JJ6), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11096879F
Description
p23/PTGES3 Monoclonal specifically detects p23/PTGES3 in Human, Mouse, Chicken, Guinea Pig, Rabbit, Yeast samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
p23/PTGES3 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
cPGESp23, Cytosolic prostaglandin E2 synthase, Hsp90 co-chaperone, P23cytosolic prostaglandin E synthase, Progesterone receptor complex p23, prostaglandin E synthase 3, prostaglandin E synthase 3 (cytosolic), TEBPEC 5.3.99.3, Telomerase-binding protein p23, unactive progesterone receptor, 23 kD | |
Mouse | |
Protein G purified | |
Membrane Trafficking and Chaperones | |
10728 | |
Human | |
Purified |
ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
JJ6 | |
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
NP_006592.3 | |
PTGES3 | |
Recombinant human full length p23 protein | |
0.1 mL | |
Primary | |
Detects approx 23kDa. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction