Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p23/PTGES3 Antibody (OTI2D2), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273186B
Description
p23/PTGES3 Monoclonal specifically detects p23/PTGES3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
p23/PTGES3 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
PTGES3 | |
Full length recombinant protein of human p23/PTGES3 (NP_006592) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2D2 | |
Western Blot, Immunohistochemistry | |
cPGESp23, Cytosolic prostaglandin E2 synthase, Hsp90 co-chaperone, P23cytosolic prostaglandin E synthase, Progesterone receptor complex p23, prostaglandin E synthase 3, prostaglandin E synthase 3 (cytosolic), TEBPEC 5.3.99.3, Telomerase-binding protein p23, unactive progesterone receptor, 23 kD | |
Mouse | |
Affinity Purified | |
Membrane Trafficking and Chaperones | |
10728.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction