Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p27/Kip1 Antibody (225501), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2256B
Description
p27/Kip1 Monoclonal antibody specifically detects p27/Kip1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOFSpecifications
p27/Kip1 | |
Monoclonal | |
Biotin | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
225501 | |
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
CDKN4, cyclin-dependent kinase inhibitor 1B, cyclin-dependent kinase inhibitor 1B (p27, Kip1), Cyclin-dependent kinase inhibitor p27, KIP1P27KIP1, MEN1B, MEN4, p27Kip1 | |
E. coli-derived recombinant human p27, Met1-Thr198, Accession # P46527 | |
0.1 mL | |
Breast Cancer, Cancer, Cell Cycle and Replication, DNA Repair, Prostate Cancer, Tumor Suppressors | |
1027 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction