Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p27/Kip1 Antibody (DCS-72.F6 + KIP1/769) - Azide and BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$646.00

Specifications

Antigen p27/Kip1
Clone DCS-72.F6 + KIP1/769
Concentration 1.0 mg/mL
Dilution Western Blot : 0.5 - 1.0 ug/ml, Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin : 0.25 - 0.5 ug/ml, Immunofluorescence : 0.5 - 1.0 ug/ml, CyTOF-ready
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP247771A
SDP
View Documents
Novus Biologicals
NBP2477710.1MG
0.1 mg
Each for $646.00
Only null left
Add to Cart
 
NBP247771B
SDP
View Documents
Novus Biologicals
NBP2477710.2MG
0.2 mg N/A N/A N/A
Description

Description

p27/Kip1 Monoclonal specifically detects p27/Kip1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence, CyTOF-ready.
Specifications

Specifications

p27/Kip1
1.0 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
Unconjugated
Mouse
Breast Cancer, Cancer, Cell Cycle and Replication, DNA Repair, Prostate Cancer, Tumor Suppressors
PBS with No Preservative
1027
Mouse recombinant p27 protein (DCS-72.F6); Recombinant human CDKN1B protein (KIP1/769)
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
25.5 kDa
DCS-72.F6 + KIP1/769
Western Blot : 0.5 - 1.0 ug/ml, Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin : 0.25 - 0.5 ug/ml, Immunofluorescence : 0.5 - 1.0 ug/ml, CyTOF-ready
Monoclonal
Purified
RUO
Human, Mouse, Rat, Monkey
CDKN4, cyclin-dependent kinase inhibitor 1B, cyclin-dependent kinase inhibitor 1B (p27, Kip1), Cyclin-dependent kinase inhibitor p27, KIP1P27KIP1, MEN1B, MEN4, p27Kip1
CDKN1B
IgG
Protein A or G purified
Recognizes a 27kDa protein, identified as the p27Kip1, a cell cycle regulatory mitotic inhibitor. Its epitope spans between aa 83-204 of p27. It is highly specific and shows no cross-reaction with other related mitotic inhibitors. p27Kip1 functions as a negative regulator of G1 progression and has been proposed to function as a possible mediator of TGF- induced G1 arrest. p27Kip1 is a candidate tumor suppressor gene. This MAb co-precipitates cdk4 in complex p27Kip1 and is excellent for staining of formalin-fixed tissues.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.