Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p27/Kip1 Antibody (KIP1/769), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP247769MFV610
Description
p27/Kip1 Monoclonal antibody specifically detects p27/Kip1 in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry, Flow (Intracellular)Specifications
p27/Kip1 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Monkey | |
Purified |
Flow Cytometry | |
KIP1/769 | |
Flow Cytometry, Flow (Intracellular) | |
CDKN4, cyclin-dependent kinase inhibitor 1B, cyclin-dependent kinase inhibitor 1B (p27, Kip1), Cyclin-dependent kinase inhibitor p27, KIP1P27KIP1, MEN1B, MEN4, p27Kip1 | |
Recombinant full-length human p27/Kip1 protein (Uniprot: P46527) | |
0.1 mL | |
Breast Cancer, Cancer, Cell Cycle and Replication, DNA Repair, Phospho Specific, Prostate Cancer, Tumor Suppressors | |
1027 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction