Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p27 Kip1 Monoclonal Antibody (DCS-72.F6), Invitrogen™

Catalog No. PIAHZ0452
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIAHZ0452 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIAHZ0452 Supplier Thermo Scientific Supplier No. AHZ0452

Mouse Monoclonal Antibody

Suggested positive controls include HT29, MAD109, PC12 cells and brain tissue.

p27 Kip1 functions as a negative regulator of G1 mouse expression and has been proposed to function as a possible mediator of TGFb induced G1 arrest. p27 Kip1 is an important regulator of cell cycle progression, a potent inhibitor of cyclin E- and cyclin A-CDK2 complexes. p27 Kip1 forms a complex with cyclin type D-CDK4 complexes and is involved in the assembly, stability, and modulation of CCND1-CDK4 complex activation. p27 Kip1 acts either as an inhibitor or an activator of cyclin type D-CDK4 complexes depending on its phosphorylation state and/or stoichometry. p27 Kip1 has a molecular weight of approximately 27kDa and the degradation of this protein, which is triggered by its CDK dependent phosphorylation and subsequent ubiquitination by SCF complexes, is required for the cellular transition from quiescence to the proliferative state.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p27 Kip1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone DCS-72.F6
Concentration 0.2 mg/mL
Conjugate Unconjugated
Gene CDKN1B
Gene Accession No. Q6SLL5, P46527, P46414
Gene Alias CDKN1B, CDN1B, Cyclin-dependent kinase inhibitor 1B, Cyclin-dependent kinase inhibitor p27, KIP1
Gene Symbols CDKN1B
Host Species Mouse
Immunogen Mouse recombit p27Kip1 protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1027, 12576, 403429, 83571
Target Species Canine, Human, Mouse, Rat
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.