Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p27/Kip1 Mouse, Clone: SX53G8, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
| Antigen | p27/Kip1 |
|---|---|
| Clone | SX53G8 |
| Dilution | Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunohistochemistry-Paraffin 0.5-1.0ug/ml |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP232889F
![]() |
Novus Biologicals
NBP2328890.02MG |
0.02 mg |
Each for $268.00
|
|
|||||
NBP232889A
![]() |
Novus Biologicals
NBP2328890.1MG |
0.1 mg |
Each for $524.00
|
|
|||||
NBP232889B
![]() |
Novus Biologicals
NBP2328890.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
p27/Kip1 Monoclonal specifically detects p27/Kip1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| p27/Kip1 | |
| Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunohistochemistry-Paraffin 0.5-1.0ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Primate | |
| P46527 | |
| 1027 | |
| Purified GST-p27 fusion protein of human origin | |
| Primary | |
| Store at 4C. |
| SX53G8 | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Breast Cancer, Cancer, Cell Cycle and Replication, DNA Repair, Phospho Specific, Prostate Cancer, Tumor Suppressors | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| CDKN4, cyclin-dependent kinase inhibitor 1B, cyclin-dependent kinase inhibitor 1B (p27, Kip1), Cyclin-dependent kinase inhibitor p27, KIP1P27KIP1, MEN1B, MEN4, p27Kip1 | |
| CDKN1B | |
| IgG1 κ | |
| Protein A purified | |
| This MAb recognizes a 27kDa protein, identified as the p27Kip1, a cell cycle regulatory mitotic inhibitor. It is highly specific and shows no cross-reaction with other related mitotic inhibitors. In Western blotting of cell lysates from 7 human breast cancer cell lines (ZR75-1, ZR75-30, MCF-7, MDAMB453, T47D, CAL51, 734B), the antibody labels a single band corresponding to p27Kip1. It functions as a negative regulator of G1 progression and has been proposed to function as a possible mediator of TGF- induced G1 arrest. p27Kip1 is a candidate tumor suppressor gene. Reportedly, low p27 expression has been associated with unfavorable prognosis in renal cell carcinoma, colon carcinoma, breast carcinomas, non-small-cell lung carcinoma, hepatocellular carcinoma, multiple myeloma, and lymph node metastases in papillary carcinoma of the thyroid, as well as a more aggressive phenotype in carcinoma of the cervix. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title