Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ p27 Kip1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111478
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111478 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111478 Supplier Invitrogen™ Supplier No. PA5111478
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PAAPLIGAPA NSEDTHLVDP KTDPSDSQTG LAEQCAGIRK RPATDDSSTQ NKRANRTEEN VSDGSPNAGS VEQTPKKPGL RRRQT Highest antigen sequence indentity to the following orthologs: Rat - 75%, Mouse - 75%.

p27 Kip1 functions as a negative regulator of G1 mouse expression and has been proposed to function as a possible mediator of TGFb induced G1 arrest. p27 Kip1 is an important regulator of cell cycle progression, a potent inhibitor of cyclin E- and cyclin A-CDK2 complexes. p27 Kip1 forms a complex with cyclin type D-CDK4 complexes and is involved in the assembly, stability, and modulation of CCND1-CDK4 complex activation. p27 Kip1 acts either as an inhibitor or an activator of cyclin type D-CDK4 complexes depending on its phosphorylation state and/or stoichometry. p27 Kip1 has a molecular weight of approximately 27kDa and the degradation of this protein, which is triggered by its CDK dependent phosphorylation and subsequent ubiquitination by SCF complexes, is required for the cellular transition from quiescence to the proliferative state.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p27 Kip1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cdkn1b
Gene Accession No. P46527
Gene Alias AA408329; AI843786; Cdki1b; CDKN1B; CDKN4; CDN1B; cyclin dependent kinase inhibitor 1B; cyclin-dependent kinase inhibitor 1B; Cyclin-dependent kinase inhibitor 1B (p27 Kip1); Cyclin-dependent kinase inhibitor 1B (p27, Kip1); cyclin-dependent kinase inhibitor p27; Cyclin-dependent kinase inhibitor p27 Kip1; cyclin-dependent kinase inhibitor p27/Kip1; KIP1; MEN1B; MEN4; p27; P27 CDKN1B; p27(KIP1); p27Kip1
Gene Symbols Cdkn1b
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human p27 Kip1. Recombinant protein control fragment (Product #RP-105121).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1027
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.