Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ p27 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552634
Description
Immunogen sequence: ITFELPDNAK QCFYEDIAQG TKCTLEFQVI TGGHYDVDCR LEDPDGKVLY KEMKKQYDSF TFTASKNGTY KFCFSNEFST F Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 99%.

Specifications
p27 | |
Polyclonal | |
Unconjugated | |
TMED7 | |
CGI-109; p24 family protein gamma-3; p24g3; p24gamma3; p27; TMED7; Transmembrane emp24 domain-containing protein 7; transmembrane emp24 protein transport domain containing 7; transmembrane p24 trafficking protein 7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51014 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.14 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y3B3 | |
TMED7 | |
Recombinant protein corresponding to Human p27. Recombinant protein control fragment (Product #RP-90226). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction