Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p27 Rat anti-Mouse, Unconjugated, Clone: SON82, MilliporeSigma™

Catalog No. MABE18151MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE18151MI 100 μg
MABE18152MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABE18151MI Supplier MilliporeSigma Supplier No. MABE1815100UG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

p27 Monoclonal antibody specifically detects p27 in Mouse samples. It is validated for Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen p27
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SON82
Conjugate Unconjugated
Gene Accession No. NP_034005
Gene Alias Cyclin-dependent kinase inhibitor 1B;Cyclin-dependent kinase inhibitor p27;p27Kip1
Gene Symbols Cdkn1b
Host Species Rat
Immunogen His-tagged full-length recombinant murine p27 protein.
Molecular Weight of Antigen 22.19 kDa calculated.
Purification Method Protein G Purified
Quantity 100 μg
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity SON82 is a rat monoclonal antibody that specifically detects mouse p27Kip1 protein.
Target Species Mouse
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.