Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2RX6 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981948
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1948 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1948 Supplier Bioss Supplier No. BS12110R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

P2rx6 encodes receptors that are members of the ligand-gated ion channel family that open in response to extracellular ATP. Each P2rx6 receptor is made up of a trimer of subunits (P2X1-7) all of which share the common structure of two transmembrane domains. The protein encoded by the P2rx6 gene belongs to the family of P2X receptors, which are ATP-gated ion channels and mediate rapid and selective permeability to cations. The P2rx6 gene is predominantly expressed in skeletal muscle, and regulated by p53. The encoded protein is associated with VE-cadherin at the adherens junctions of human umbilical vein endothelial cells. Alternative splicing results in multiple transcript variants of P2rx6.

Specifications

Antigen P2RX6
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene P2RX6
Gene Accession No. O15547
Gene Alias ATP receptor; P2rx6; P2RXL1; P2X purinoceptor 6; P2X6; P2X6 receptor; P2XM; Purinergic receptor; purinergic receptor P2X 6; purinergic receptor P2X, ligand gated ion channel, 6; purinergic receptor P2X, ligand-gated ion channel, 6; purinergic receptor P2X6; Purinergic receptor P2X-like 1; purinergic receptor P2X-like 1 orphan receptor; purinergic receptor P2X-like 1, orphan receptor; purinoceptor P2X6; purinoreceptor P2X6; Rnap2x6; skeletal muscle-expressed P2X
Gene Symbols P2RX6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human P2RX6.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9127
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.