Learn More
P2RX7 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | P2RX7 |
|---|---|
| Concentration | 300 μg/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-8638
![]()
|
Proteintech Group Inc
282071AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The P2RX7 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human P2RX7. This antibody recognizes human, mouse antigen. The P2RX7 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| P2RX7 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Human, Mouse | |
| IgG |
| 300 μg/mL | |
| Polyclonal | |
| Rabbit | |
| 5027 | |
| Primary |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.