Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2RY13 Mouse anti-Human, Clone:3E8C12, Abnova™

Catalog No. 89333163 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-163 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-163 Supplier Abnova Corporation Supplier No. MAB16713
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human P2RY13.

The product of this gene belongs to the family of G-protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor is activated by ADP. [provided by RefSeq]

Specifications

Antigen P2RY13
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 3E8C12
Conjugate Unconjugated
Description purinergic receptor P2Y, G-protein coupled, 13
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry
Immunocytochemistry
Flow Cytometry (1:200-1:400)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene P2RY13
Gene Alias FKSG77, GPCR1, GPR86, GPR94, P2Y13, SP174
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 1-49 of human P2RY13 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 53829
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.