Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P2X1/P2RX1 Antibody (551820) [Allophycocyanin], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10533A
Description
P2X1/P2RX1 Monoclonal antibody specifically detects P2X1/P2RX1 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
P2X1/P2RX1 | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
551820 | |
Flow Cytometry, CyTOF-ready | |
ATP receptor, P2X purinoceptor 1, P2X receptor, subunit 1, P2X1P2X1 receptor, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 1 | |
Human embryonic kidney cell, HEK293-derived transfected with human P2X1/P2RX1, Accession # P51575 | |
0.1 mL | |
Signal Transduction | |
5023 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction