Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ P2X1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144864
Description
Immunogen sequence: LFIKNSISFP RFKVNRRNLV EEVNAAHMKT CLFHKTLHPL CPVFQLGYVV QESGQNFSTL AEKGGVV Antigen sequence identity: Mouse 84%; Rat 82%.
The product of this gene belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel with relatively high calcium permeability. Binding to ATP mediates synaptic transmission between neurons and from neurons to smooth muscle, being responsible, for example, for sympathetic vasoconstriction in small arteries, arterioles and vas deferens. Mouse studies suggest that this receptor is essential for normal male reproductive function. It is possible that the development of selective antagonists for this receptor may provide an effective non-hormonal male contraceptive pill.
Specifications
P2X1 | |
Polyclonal | |
Unconjugated | |
P2RX1 | |
AI323649; ATP receptor; BB122383; P2RX1; P2x; P2X purinoceptor 1; P2X receptor, subunit 1; P2X1; P2X1 receptor; P2XMR; Pdcd3; programmed cell death 3; Purinergic receptor; purinergic receptor P2X 1; purinergic receptor P2X, ligand gated ion channel, 1; purinergic receptor P2X, ligand-gated ion channel, 1; purinergic receptor P2X1; purinergic receptor P2X1-1; purinergic receptor P2X1-2; RP-2; RP-2 protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
5023 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P51575 | |
P2RX1 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 187-253. Recombinant protein control fragment (Product #RP-110019). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction