Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P2X1 Polyclonal Antibody
P2X1 Polyclonal Antibody
Supplier: Thermo Scientific PA5103994
Description
P2X1 Polyclonal Antibody for Western Blot, ICC/IF

Specifications
P2X1 | |
Polyclonal | |
Unconjugated | |
P2RX1 | |
AI323649; ATP receptor; BB122383; P2RX1; P2x; P2X purinoceptor 1; P2X receptor, subunit 1; P2X1; P2X1 receptor; P2XMR; Pdcd3; programmed cell death 3; Purinergic receptor; purinergic receptor P2X 1; purinergic receptor P2X, ligand gated ion channel, 1; purinergic receptor P2X, ligand-gated ion channel, 1; purinergic receptor P2X1; purinergic receptor P2X1-1; purinergic receptor P2X1-2; RP-2; RP-2 protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
18436, 25505, 5023 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P47824, P51575, P51576 | |
P2RX1 | |
A synthesized peptide derived from human P2RX1(Accession P51575), corresponding to amino acid residues T256-H306. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction