Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ P2X2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531318
Description
Recommended positive controls: A549, HepG2, mouse brain. Predicted reactivity: Mouse (81%), Rat (80%), Bovine (86%), Guinea pig (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
P2rx2 are members of the ligand-gated ion channel family that open in response to extracellular ATP. Each receptor is made up of a trimer of subunits (P2X1-7) all of which share the common structure of two transmembrane domains. P2rx2 functions as a ligand-gated ion channel and binding to ATP mediates synaptic transmission between neurons and from neurons to smooth muscle. Six transcript variants encoding six distinct isoforms have been identified for P2rx2. P2rx2 is involved in a variety of cellular responses, such as excitatory postsynaptic responses in sensory neurons, neuromuscular junctions (NMJ) formation, hearing, perception of taste and peristalsis. In the inner ear, P2rx2 regulates sound transduction and auditory neurotransmission, outer hair cell electromotility, inner ear gap junctions, and K(+) recycling. P2rx2 also mediates synaptic transmission between neurons and from neurons to smooth muscle. Diseases associated with P2RX2 include Deafness, Autosomal Dominant 41 and Dfna41 Nonsyndromic Hearing Loss And Deafness.
Specifications
P2X2 | |
Polyclonal | |
Unconjugated | |
P2RX2 | |
ATP receptor; DFNA41; MGC129601; P2RX2; P2X purinoceptor 2; P2X Receptor, subunit 2; P2X2; P2X2 purinoceptor; P2X2 receptor splice variant P2X2-1; P2X2 receptor splice variant P2X2-2; P2X2 receptor splice variant P2X2-3; P2X2a; Purinergic receptor; purinergic receptor P2X 2; purinergic receptor P2X, ligand gated ion channel, 2; purinergic receptor P2X, ligand-gated ion channel, 2; purinergic receptor P2X2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22953, 231602 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8K3P1, Q9UBL9 | |
P2RX2 | |
Recombinant fragment corresponding to a region within amino acids 8 and 302 of Human P2X2. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction