Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ P2X3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534067
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset (100%); Bat, Rabbit, Opossum (93%); Mouse, Rat, Hamster, Elephant, Panda, Bovine, Dog, Horse (87%). Antigen retrieval requires Proteinase K treatment.
The product of this gene belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel and may transduce ATP-evoked nociceptor activation. Mouse studies suggest that this receptor is important for peripheral pain responses, and also participates in pathways controlling urinary bladder volume reflexes. It is possible that the development of selective antagonists for this receptor may have a therapeutic potential in pain relief and in the treatment of disorders of urine storage.
Specifications
P2X3 | |
Polyclonal | |
Unconjugated | |
P2rx3 | |
4930513E20Rik; ATP receptor; MGC129956; P2RX3; P2X purinoceptor 3; p2X receptor 3; P2X receptor, subunit 3; P2X3; p2X3 purinoceptor; p2xr3; Purinergic receptor; purinergic receptor P2X 3; purinergic receptor P2X, ligand gated ion channel, 3; purinergic receptor P2X, ligand-gated ion channel, 3; purinergic receptor P2X3; purinoceptor P2X3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5024 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P56373 | |
P2rx3 | |
Synthetic 15 amino acid peptide from C-terminal cytoplasmic domain of human P2RX3 / P2X3. | |
50 μg | |
Primary | |
Human, Monkey | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction