Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ P2X4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576989
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576989 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576989 Supplier Invitrogen™ Supplier No. PA576989
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

The product of this gene belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel with high calcium permeability. The main pharmacological distinction between the members of the purinoceptor family is the relative sensitivity to the antagonists suramin and PPADS. The product of this gene has the lowest sensitivity for these antagonists. Multiple alternatively spliced transcript variants have been identified for this gene although their full-length natures have not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P2X4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene P2RX4
Gene Accession No. Q99571, Q9JJX6
Gene Alias AI504491; ATP receptor; ATP-gated cation channel protein; AW555605; D5Ertd444e; ionotropic purinergic receptor; P2 purinergic receptor subtype; P2RX4; P2X purinoceptor 4; P2X receptor, subunit 4; P2X4; P2X4 purinoceptor; P2x4 receptor; P2X4R; Purinergic receptor; purinergic receptor P2X 4; purinergic receptor P2X, ligand gated ion channel, 4; purinergic receptor P2X, ligand-gated ion channel 4; purinergic receptor P2X, ligand-gated ion channel, 4; purinergic receptor P2X4; purinergic receptor P2X4 isoform a; purinergic receptor P2X4 subunit variant 1; purinergic receptor P2X4 subunit variant 2; purinergic receptor P2x4 subunit variant e; purinoceptor P2X4; putative partial extracellular loop region
Gene Symbols P2RX4
Host Species Rabbit
Immunogen Recombinant full length Human P2X4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18438, 5025
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.