Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ P2X4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA593144
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA593144 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA593144 Supplier Invitrogen™ Supplier No. PA593144
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QETDSVVSSV TTKVKGVAVT NTSKLGFRIW DVADYVIPAQ EENSLFVMTN VILTMNQTQG LCPEIPDATT VCKSDASCTA GSAGTHSNGV STGRCVAFNG SVKTCEVAAW CPVEDDTHVP QPAFLKAAEN FTLLVKNNIW YPKFNFSKRN ILPNITTTYL KSCIYDAKTD PFCPIFRLGK IVENAGHSFQ DMAVEGGIMG IQVNWDCNLD RAASLCLPRY SFRRLDTRDV EHNVSPGYNF RFAKYYRDLA GNEQRTLIKA YGIRFDIIVF GKAGKFDIIP TMIN; Positive Samples: 22Rv1, Mouse kidney, Mouse lung; Cellular Location: Membrane, Multi-pass membrane protein.

The product of this gene belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel with high calcium permeability. The main pharmacological distinction between the members of the purinoceptor family is the relative sensitivity to the antagonists suramin and PPADS. The product of this gene has the lowest sensitivity for these antagonists. Multiple alternatively spliced transcript variants have been identified for this gene although their full-length natures have not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P2X4
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene P2RX4
Gene Accession No. Q99571
Gene Alias AI504491; ATP receptor; ATP-gated cation channel protein; AW555605; D5Ertd444e; ionotropic purinergic receptor; P2 purinergic receptor subtype; P2RX4; P2X purinoceptor 4; P2X receptor, subunit 4; P2X4; P2X4 purinoceptor; P2x4 receptor; P2X4R; Purinergic receptor; purinergic receptor P2X 4; purinergic receptor P2X, ligand gated ion channel, 4; purinergic receptor P2X, ligand-gated ion channel 4; purinergic receptor P2X, ligand-gated ion channel, 4; purinergic receptor P2X4; purinergic receptor P2X4 isoform a; purinergic receptor P2X4 subunit variant 1; purinergic receptor P2X4 subunit variant 2; purinergic receptor P2x4 subunit variant e; purinoceptor P2X4; putative partial extracellular loop region
Gene Symbols P2RX4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 55-338 of human P2RX4 (NP_0025512).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5025
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.