Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P2X5/P2RX5 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298199DL594
Description
P2X5/P2RX5 Polyclonal specifically detects P2X5/P2RX5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
P2X5/P2RX5 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
ATP receptor, ATP receptor subunit, ionotropic ATP receptor P2X5, LRH-1, MGC47755, P2X purinoceptor 5, P2X5lymphoid-restricted histocompatibility antigen-1, P2X5R, Purinergic receptor, purinergic receptor P2X ligand gated ion channel 5, purinergic receptor P2X, ligand-gated ion channel, 5 | |
Produced in rabbits immunized with E. coli-derived Human P2X5/P2RX5 fragment. | |
0.1 mL | |
Signal Transduction | |
5026 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction