Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P2X5/P2RX5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP256419
Description
P2X5/P2RX5 Polyclonal specifically detects P2X5/P2RX5 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
P2X5/P2RX5 | |
Polyclonal | |
Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
ATP receptor, ATP receptor subunit, ionotropic ATP receptor P2X5, LRH-1, MGC47755, P2X purinoceptor 5, P2X5lymphoid-restricted histocompatibility antigen-1, P2X5R, Purinergic receptor, purinergic receptor P2X ligand gated ion channel 5, purinergic receptor P2X, ligand-gated ion channel, 5 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
P2RX5 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TNLIVTPNQRQNVCAENEGIPDGACSKDSDCHAGEAVTAGNGVKTGRCLRRENLARGTCEIFAWCPLETSSRPEEPFLKE | |
100 μL | |
Signal Transduction | |
5026 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction