Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ P2X5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566958
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566958 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566958 Supplier Invitrogen™ Supplier No. PA566958
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TNLIVTPNQRQ NVCAENEGIP DGACSKDSDC HAGEAVTAGN GVKTGRCLRR ENLARGTCEI FAWCPLETSS RPEEPFLKE Highest antigen sequence identity to the following orthologs - mouse 74%, rat 74%.

The P2X5 gene encodes a protein that is a member of the purinergic receptor family, P2X receptors. These receptors are ligand-gated ion channels activated by extracellular ATP. The P2X5 protein is primarily involved in the regulation of skeletal muscle, immune response, and sensory neuron functions. Structurally, P2X5 features two transmembrane domains, a large extracellular loop, and intracellular N- and C-termini. P2X5 is known to form both homomeric and heteromeric channels, contributing to its functional diversity in various tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P2X5
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene P2RX5
Gene Accession No. Q93086
Gene Alias AI852046; ATP receptor; ATP receptor subunit; ionotropic ATP receptor P2X5; LRH-1; lymphoid-restricted histocompatibility antigen-1; membrane magnesium transporter 2; membrane magnesium transporter 2 {ECO:0000250; MGC47755; Mmgt2; mmgt2 {ECO:0000250; P2 purinergic receptor subtype; P2RX5; P2X purinoceptor 5; P2X5; P2X5 purinoceptor; P2x5 receptor; P2X5R; Purinergic receptor; purinergic receptor P2X 5; purinergic receptor P2X, ligand gated ion channel, 5; purinergic receptor P2X, ligand-gated ion channel, 5; purinergic receptor P2X5-1; purinergic receptor P2X5-2; RGD1311260; UniProtKB:Q8R3L0}
Gene Symbols P2RX5
Host Species Rabbit
Immunogen Recombinant Human P2X5. Recombinant protein control fragment (Product #RP-107603).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5026
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.