Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ P2X5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579778
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Brain Tissue. IHC: Mouse Brain Tissue, Rat Brain Tissue.
Mmgt2 mediates Mg(2+) transport and may provide regulated pathways for Mg(2+) transport in the Golgi and post-Golgi organelles of epithelium-derived cells. Magnesium is an essential metal, but few selective transporters have been identified. The two separate gene products, Mmgt1 and Mmgt2, comprise the family that we have termed membrane Mg(2+) transporters (MMgTs), and the proteins reside in the membrane and mediate Mg(2+) transport. The transport of Mg(2+) is saturable and studies have identified that Mmgt2 fusion proteins reside in the Golgi complex and post-Golgi vesicles, including the early endosomes in COS-7 cells transfected with the respective tagged constructs. Among related pathways for Mmgt2 are transport and binding events, in particular, transport of glucose and other sugars, bile salts and organic acids, metal ions and amine compounds.
Specifications
P2X5 | |
Polyclonal | |
Unconjugated | |
P2RX5 | |
AI852046; ATP receptor; ATP receptor subunit; ionotropic ATP receptor P2X5; LRH-1; lymphoid-restricted histocompatibility antigen-1; membrane magnesium transporter 2; membrane magnesium transporter 2 {ECO:0000250; MGC47755; Mmgt2; mmgt2 {ECO:0000250; P2 purinergic receptor subtype; P2RX5; P2X purinoceptor 5; P2X5; P2X5 purinoceptor; P2x5 receptor; P2X5R; Purinergic receptor; purinergic receptor P2X 5; purinergic receptor P2X, ligand gated ion channel, 5; purinergic receptor P2X, ligand-gated ion channel, 5; purinergic receptor P2X5-1; purinergic receptor P2X5-2; RGD1311260; UniProtKB:Q8R3L0} | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
113995, 216829, 303211, 5026, 94045 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5 mg BSA and 0.05 mg sodium azide | |
P51578, Q6P719, Q8R3L0, Q93086 | |
Mmgt2, P2RX5 | |
E.coli-derived human P2X5 recombinant protein (Position: D333-T422). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction