Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ P2X6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587659
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587659 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587659 Supplier Invitrogen™ Supplier No. PA587659
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WGLLDYKTEK WALLAKKGYQ ERDLEPQFSI ITKLKGVSVT QIKELGNRLW DVADFVKPPQ GENVFFLVTN FLVTPAQVQG RCPEHPSVPL ANCWVDEDCP EGEGGTHSHG VKTGQCVVFN GTHRTCEIWS WCPVESGVVP SRPLLAQAQN FTLFIKNTVT FSKFNFSKSN ALETWDPTYF KHCRYEPQFS PYCPVFRIGD LVAKAGGTFE DLALLGGSVG IRVHWDCDLD TGDSGCWPHY SFQLQEKSYN FRTATHWWEQ P; Positive Samples: Mouse pancreas; Cellular Location: Membrane, Multi-pass membrane protein.

P2rx6 encodes receptors that are members of the ligand-gated ion channel family that open in response to extracellular ATP. Each P2rx6 receptor is made up of a trimer of subunits (P2X1-7) all of which share the common structure of two transmembrane domains. The protein encoded by the P2rx6 gene belongs to the family of P2X receptors, which are ATP-gated ion channels and mediate rapid and selective permeability to cations. The P2rx6 gene is predominantly expressed in skeletal muscle, and regulated by p53. The encoded protein is associated with VE-cadherin at the adherens junctions of human umbilical vein endothelial cells. Alternative splicing results in multiple transcript variants of P2rx6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P2X6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene P2RX6
Gene Accession No. O54803
Gene Alias ATP receptor; P2RX6; P2RXL1; P2X purinoceptor 6; P2x6; P2X6 receptor; P2XM; Purinergic receptor; purinergic receptor P2X 6; purinergic receptor P2X, ligand gated ion channel, 6; purinergic receptor P2X, ligand-gated ion channel, 6; purinergic receptor P2X6; Purinergic receptor P2X-like 1; purinergic receptor P2X-like 1 orphan receptor; purinergic receptor P2X-like 1, orphan receptor; purinoceptor P2X6; purinoreceptor P2X6; Rnap2x6; skeletal muscle-expressed P2X
Gene Symbols P2RX6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 20-280 of human P2RX6 (NP_0011530261).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18440
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.