Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ P2X7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587959
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587959 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587959 Supplier Invitrogen™ Supplier No. PA587959
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: A375, HT-29, U-251MG, LO2, Mouse heart, Rat liver; Cellular Location: Cell membrane, Multi-pass membrane protein Immunogen sequence: SDKLYQRKEP VISSVHTKVK GIAEVKEEIV ENGVKKLVHS VFDTADYTFP LQGNSFFVMT NFLKTEGQEQ RLCPEYPTRR TLCSSDRGCK KGWMDPQSKG IQTGRCVVYE GNQKTCEVSA WCPIEAVEEA PRPALLNSAE NFTVLIKNNI DFPGHNYTTR NILPGLNITC TFHKTQNPQC PIFRLGDIFR ETGDNFSDVA IQGGIMGIEI YWDCNLDRWF HHCRPKYSFR RLDDKTTNVS LYPGYNFRYA KYYKENNVEK RTLIKVFGIR FDILVFGTGG KFDIIQLV.

The product of this gene belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel and is responsible for ATP-dependent lysis of macrophages through the formation of membrane pores permeable to large molecules. Activation of this nuclear receptor by ATP in the cytoplasm may be a mechanism by which cellular activity can be coupled to changes in gene expression. Multiple alternatively spliced variants have been identified, most of which fit nonsense-mediated decay criteria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P2X7
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene P2RX7
Gene Accession No. Q64663, Q99572, Q9Z1M0
Gene Alias AI467586; ATP receptor; MGC20089; P2rx7; P2X purinoceptor 7; p2X receptor 7; P2X(7); P2X7; P2X7 purinoceptor; P2X7 receptor; P2X7R; P2Z; P2Z receptor; Purinergic receptor; purinergic receptor P2X 7; purinergic receptor P2X, ligand gated ion channel, 7; purinergic receptor P2X, ligand-gated ion channel, 7; purinergic receptor P2X7; purinergic receptor P2X7 variant A
Gene Symbols P2RX7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 47-334 of human P2RX7 (NP_0025533).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18439, 29665, 5027
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.