Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2X7 Polyclonal Antibody, Huabio

Catalog No. 502544688
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4688 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4688 Supplier HUABIO RESEARCH INC Supplier No. ER190199
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

P2X7 Polyclonal antibody specifically detects P2X7 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen P2X7
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene P2RX7
Gene Accession No. Q64663, Q99572, Q9Z1M0
Gene Alias AI467586; ATP receptor; MGC20089; P2rx7; P2X purinoceptor 7; p2X receptor 7; P2X(7); P2X7; P2X7 purinoceptor; P2X7 receptor; P2X7R; P2Z; P2Z receptor; Purinergic receptor; purinergic receptor P2X 7; purinergic receptor P2X, ligand gated ion channel, 7; purinergic receptor P2X, ligand-gated ion channel, 7; purinergic receptor P2X7; purinergic receptor P2X7 variant A
Host Species Rabbit
Immunogen Synthetic peptide corresponding to C terminal Rat P2X7.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18439, 29665, 5027
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.