Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ P2Y1 Monoclonal Antibody (4G5B7)
GREENER_CHOICE

Catalog No. PIMA531815
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531815 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531815 Supplier Invitrogen™ Supplier No. MA531815
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Amino acid sequence of the immunogen is proprietary. This antibody has been tested in indirect ELISA.

P2Y1 is a Purinergic Receptor that causes a change in platelet shape and potentially leads to platelet aggregation by binding ADP, which results in the mobilization of intracellular calcium ions via activation of phospholipase C. This receptor also mediates the release of the relaxing factor nitric oxide and may be involved in the modulation of neuronal signal transmission. P2Y1 has been reported to be expressed in tissues throughout the body, particularly brain. ESTs have been isolated from human embryo, lung, and placenta libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P2Y1
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4G5B7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene P2ry1
Gene Accession No. P47900
Gene Alias ADP receptor; ATP receptor; P2 purinoceptor subtype Y1; P2 purinoreceptor subclass 2Y; P2RY1; P2y; P2Y ATP receptor 1; P2Y purinoceptor 1; P2Y1; P2Y1 receptor; p2y1r; p2y-1-r; platelet ADP receptor; Purinergic receptor; purinergic receptor P2Y, G-protein coupled 1; purinergic receptor P2Y, G-protein coupled, 1; purinergic receptor P2Y1; Purinoceptor p2y1; Purinoceptor p2yr1
Gene Symbols P2ry1
Host Species Mouse
Immunogen Purified recombinant fragment of human P2RY1 expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5028
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.