Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ P2Y1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534069
Description
Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Gibbon, Monkey, Mouse (94%) Marmoset, Rat, Hamster, Bovine (89%) Rabbit, Pig (83%).
P2Y1 is a Purinergic Receptor that causes a change in platelet shape and potentially leads to platelet aggregation by binding ADP, which results in the mobilization of intracellular calcium ions via activation of phospholipase C. This receptor also mediates the release of the relaxing factor nitric oxide and may be involved in the modulation of neuronal signal transmission. P2Y1 has been reported to be expressed in tissues throughout the body, particularly brain. ESTs have been isolated from human embryo, lung, and placenta libraries.
Specifications
| P2Y1 | |
| Polyclonal | |
| Unconjugated | |
| P2ry1 | |
| ADP receptor; ATP receptor; P2 purinoceptor subtype Y1; P2 purinoreceptor subclass 2Y; P2RY1; P2y; P2Y ATP receptor 1; P2Y purinoceptor 1; P2Y1; P2Y1 receptor; p2y1r; p2y-1-r; platelet ADP receptor; Purinergic receptor; purinergic receptor P2Y, G-protein coupled 1; purinergic receptor P2Y, G-protein coupled, 1; purinergic receptor P2Y1; Purinoceptor p2y1; Purinoceptor p2yr1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5028 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P47900 | |
| P2ry1 | |
| Synthetic 18 amino acid peptide from N-terminal extracellular domain of human P2RY1 / P2Y1. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction