Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P2Y10/P2RY10 Antibody (639329), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10158APCCY7
Description
P2Y10/P2RY10 Monoclonal antibody specifically detects P2Y10/P2RY10 in Human samples. It is validated for Flow CytometrySpecifications
P2Y10/P2RY10 | |
Monoclonal | |
APC-Cyanine7 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
639329 | |
Flow Cytometry | |
G-protein coupled purinergic receptor P2Y10, P2Y purinoceptor 10, P2Y10P2Y-like receptor, purinergic receptor P2Y, G-protein coupled, 10, putative P2Y purinoceptor 10 | |
NS0 mouse myeloma cell line transfected with human P2RY10, Accession # O00398 | |
0.1 mL | |
Apoptosis, Cytokine Research, GPCR, Plasma Membrane Markers | |
27334 | |
Store at 4C in the dark. Do not freeze. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction