Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ P2Y10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534075
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534075 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534075 Supplier Invitrogen™ Supplier No. PA534075
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Hamster (100%) Gibbon, Mouse, Rat, Panda, Bat, Bovine, Rabbit, Horse (94%).

P2Y10 belongs to the family of G-protein coupled receptors, that are preferentially activated by adenosine and uridine nucleotides. Two alternatively spliced transcript variants encoding the same protein isoform have been found for this gene. P2Y10 has been reported to be expressed in lymphoid cells and in monocytes derived from dendritic cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P2Y10
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene P2RY10
Gene Accession No. O00398
Gene Alias 5830408N17Rik; G-protein coupled purinergic receptor P2Y10; LYPSR2; P2RY10; p2Y purinoceptor 10; P2Y10; P2Y-like receptor; purinergic receptor P2Y, G-protein coupled 10; purinergic receptor P2Y, G-protein coupled, 10; purinergic receptor P2Y10; putative P2Y purinoceptor 10; RGD1561912; RP3-333E23.1
Gene Symbols P2RY10
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from C-terminus of human P2RY10 / P2Y10.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27334
Target Species Human, Monkey, Hamster
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.