Learn More
Invitrogen™ P2Y10 Polyclonal Antibody

Description
This target displays homology in the following species: Human: 100%; Mouse: 86%; Rat: 100%.
Specifications
Specifications
| Antigen | P2Y10 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | P2RY10 |
| Gene Accession No. | O00398 |
| Gene Alias | 5830408N17Rik; G-protein coupled purinergic receptor P2Y10; LYPSR2; P2RY10; p2Y purinoceptor 10; P2Y10; P2Y-like receptor; purinergic receptor P2Y, G-protein coupled 10; purinergic receptor P2Y, G-protein coupled, 10; purinergic receptor P2Y10; putative P2Y purinoceptor 10; RGD1561912; RP3-333E23.1 |
| Gene Symbols | P2RY10 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.