Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P2Y11/P2RY11 Antibody (505214), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB9305H
Description
P2Y11/P2RY11 Monoclonal antibody specifically detects P2Y11/P2RY11 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| P2Y11/P2RY11 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| 0.1 mL | |
| Apoptosis, Cytokine Research, GPCR, Plasma Membrane Markers | |
| 5032 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 505214 | |
| Flow Cytometry, CyTOF-ready | |
| P2Y11P2Y purinoceptor 11, purinergic receptor P2Y, G-protein coupled, 11, purinergic receptor P2Y11 | |
| NS0 mouse myeloma cell line transfected with human P2Y11/P2RY11, Accession # Q96G91 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction