Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2Y11 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA120615
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA120615 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA120615 Supplier Thermo Scientific Supplier No. PA120615

Rabbit Polyclonal Antibody

Positive control: Dendritic cells

P2Y11 belongs to the family of G protein coupled receptors, and is coupled to the stimulation of the phosphoinositide and adenylyl cyclase pathways and behaves as a selective purinoceptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P2Y11
Applications Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene P2RY11
Gene Accession No. Q96G91
Gene Alias P2RY11; P2Y purinoceptor 11; P2Y11; purinergic receptor P2Y, G-protein coupled, 11; purinergic receptor P2Y11
Gene Symbols P2RY11
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from 3rd extracellular domain of human P2Y11
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5032
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.