Learn More
Description
Specifications
Specifications
| Antigen | P2Y12/P2RY12 |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Clone | 1C2A9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | ADPG-R, Gi-coupled ADP receptor HORK3, HORK3P2Y12 platelet ADP receptor, P2T(AC), P2Y purinoceptor 12, P2Y(AC), P2Y(ADP), P2Y(cyc), P2Y12ADP-glucose receptor, purinergic receptor P2RY12, purinergic receptor P2Y, G-protein coupled, 12, putative G-protein coupled receptor, SP1999G-protein coupled receptor SP1999 |
| Gene Symbols | P2RY12 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
