Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2Y12/P2RY12 Antibody (1C2A9) - BSA Free, Novus Biologicals™
SDP

Catalog No. p-200080729 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-61749 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-61749 Supplier Novus Biologicals Supplier No. NBP261749
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 1 publication

P2Y12/P2RY12 Monoclonal specifically detects P2Y12/P2RY12 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen P2Y12/P2RY12
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 1C2A9
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
Formulation PBS with 0.05% Sodium Azide
Gene Alias ADPG-R, Gi-coupled ADP receptor HORK3, HORK3P2Y12 platelet ADP receptor, P2T(AC), P2Y purinoceptor 12, P2Y(AC), P2Y(ADP), P2Y(cyc), P2Y12ADP-glucose receptor, purinergic receptor P2RY12, purinergic receptor P2Y, G-protein coupled, 12, putative G-protein coupled receptor, SP1999G-protein coupled receptor SP1999
Gene Symbols P2RY12
Host Species Mouse
Immunogen Purified recombinant fragment of human P2RY12 (AA: extra mix) expressed in E. Coli.
Molecular Weight of Antigen 39.4 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 64805
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.