Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ P2Y12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534080
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534080 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534080 Supplier Invitrogen™ Supplier No. PA534080
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Panda, Dog, Horse, Rabbit (100%) Bat, Pig, Platypus (94%).

P2Y12 belongs to the family of G-protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor is involved in platelets aggregation, and is a potential target for the treatment of thromboembolisms and other clotting disorders. Two transcript variants encoding the same isoform have been identified for this gene. Expression of P2Y12 has been reported in platelets, spinal cord, and many brain regions. ESTs for P2Y12 have been isolated from B-cell/lung/testis, brain, embryo, prostate, eye, kidney carcinoma, and colon carcinoma libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P2Y12
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene P2ry12
Gene Accession No. Q9H244
Gene Alias 2900079B22Rik; 4921504D23Rik; ADP-glucose receptor; ADPG-R; BDPLT8; Gi-coupled ADP receptor HORK3; G-protein coupled receptor SP1999; HORK3; P2RY12; P2T(AC); P2Y purinoceptor 12; P2Y(12)R; P2Y(AC); P2Y(ADP); P2Y(cyc); P2y12; P2Y12 platelet ADP receptor; P2YR12; purinergic receptor P2RY12; purinergic receptor P2Y, G-protein coupled 12; purinergic receptor P2Y, G-protein coupled, 12; purinergic receptor P2Y12; putative G-protein coupled receptor; SP1999
Gene Symbols P2ry12
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from 2nd extracellular domain of human P2RY12 / P2Y12.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100345006, 100630046, 442958, 64805
Target Species Human, Canine, Rabbit, Monkey, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.