Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2Y12 Receptor, Rabbit anti-Rat, Human, Polyclonal, Millipore Sigma™

Catalog No. ABS1487100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS1487100 100 μg
ABS148725UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABS1487100 Supplier MilliporeSigma Supplier No. ABS1487100UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-P2Y12 Receptor, Cat. No. ABS1487, is a rabbit polyclonal antibody that detects P2Y purinoceptor 12 and has been tested for Immunohistochemistry (Paraffin) and Western Blotting.

P2Y purinoceptor 12 (UniProt: Q9H244; also known as P2Y12, ADP-glucose receptor, ADPG-R, P2T(AC), P2Y(AC), P2Y(cyc), P2Y12 platelet ADP receptor, P2Y(ADP), SP1999) is encoded by the P2RY12 (also known as HORK3) gene (Gene ID: 64805) in human. P2Y12 is a multi-pass metabotropic receptor that is highly expressed in the platelets. Lower expression levels have been observed in the brain, lung, and adrenal glands. P2Y12 serves as a receptor for ADP and ATP coupled to G-proteins that inhibit the adenylyl cyclase second messenger system. It is not activated by UDP and UTP. It is required for normal platelet aggregation and blood coagulation. It is the predominant receptor involved in the ADP-stimulated activation of the glycoprotein IIb/IIIa receptor. Activation of the glycoprotein IIb/IIIa receptor results in enhanced platelet degranulation and thromboxane production, and prolonged platelet aggregation. P2Y12 receptor has also been reported to be expressed on microglia and is required to neuropathic pain following peripheral nerve injury. P2Y12 has seven transmembrane, four cytoplasmic, and four extracellular domains. The transmembrane helices are somewhat tilted and/or kinked. Agonist binding promotes a conformation change in the extracellular loops that leads to an inward movement of the transmembrane helices. Mutations in P2RY12 gene have been linked to bleeding disorders resulting from severe impairment of platelet response to ADP resulting in defective platelet aggregation. (Ref.: Burnstock G. (2009). Curr. Pharm. Des. 15(15);1717-35; Damman, P., et al. (2012). J. Thromb. Thrombolysis. 33(2); 143-153).
TRUSTED_SUSTAINABILITY

Specifications

Antigen P2Y12 Receptor
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry (Paraffin) Analysis: A 1:250-1,000 dilution from a representative lot detected P2Y12 Receptor in human cerebral cortex and human tonsil tissue sections.Western Blotting Analysis: 1μg/mL from a representative lot detected P2Y12 Receptor in human platelet cell lysate.
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias P2Y purinoceptor 12;ADP-glucose receptor;ADPG-R;P2T(AC);P2Y(AC);P2Y(cyc);P2Y12 platelet ADP receptor;P2Y(ADP);SP1999
Gene Symbols P2RY12;HORK3
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 13 amino acids from the internal region of human P2Y purinoreceptor 12.
Molecular Weight of Antigen ∽50 kDa observed; 39.44 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_073625
Test Specificity This rabbit polyclonal antibody specifically detects P2Y12 receptor. It targets an epitope with in 13 amino acids from the extracellular domain.
Target Species Rat, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.