Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ P2Y12 Recombinant Rabbit Monoclonal Antibody (JE63-23)
GREENER_CHOICE

Catalog No. PIMA544796
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544796 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544796 Supplier Invitrogen™ Supplier No. MA544796
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

P2Y12 Recombinant Monoclonal Antibody for Western Blot

P2Y12 belongs to the family of G-protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor is involved in platelets aggregation, and is a potential target for the treatment of thromboembolisms and other clotting disorders. Two transcript variants encoding the same isoform have been identified for this gene. Expression of P2Y12 has been reported in platelets, spinal cord, and many brain regions. ESTs for P2Y12 have been isolated from B-cell/lung/testis, brain, embryo, prostate, eye, kidney carcinoma, and colon carcinoma libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P2Y12
Applications Western Blot
Classification Recombinant Monoclonal
Clone JE63-23
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene P2ry12
Gene Accession No. Q9CPV9, Q9H244
Gene Alias 2900079B22Rik; 4921504D23Rik; ADP-glucose receptor; ADPG-R; BDPLT8; Gi-coupled ADP receptor HORK3; G-protein coupled receptor SP1999; HORK3; P2RY12; P2T(AC); P2Y purinoceptor 12; P2Y(12)R; P2Y(AC); P2Y(ADP); P2Y(cyc); P2y12; P2Y12 platelet ADP receptor; P2YR12; purinergic receptor P2RY12; purinergic receptor P2Y, G-protein coupled 12; purinergic receptor P2Y, G-protein coupled, 12; purinergic receptor P2Y12; putative G-protein coupled receptor; SP1999
Gene Symbols P2ry12
Host Species Rabbit
Immunogen Recombinant protein within human P2Y12 aa 101-342/342.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64805, 70839
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.