Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2Y13/P2RY13/GPR86 Antibody (3E8C12) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP252527 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP252527 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP252527 Supplier Novus Biologicals Supplier No. NBP252527
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

P2Y13/P2RY13/GPR86 Monoclonal specifically detects P2Y13/P2RY13/GPR86 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.

Specifications

Antigen P2Y13/P2RY13/GPR86
Applications Western Blot, Flow Cytometry, ELISA, CyTOF
Classification Monoclonal
Clone 3E8C12
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready
Formulation PBS with 0.05% Sodium Azide
Gene Alias G protein-coupled receptor 86, GPR86GPCR1, GPR94, G-protein coupled receptor 86, G-protein coupled receptor 94, P2Y purinoceptor 13, P2Y13FKSG77, purinergic receptor P2Y, G-protein coupled, 13, SP174
Gene Symbols P2RY13
Host Species Mouse
Immunogen Purified recombinant fragment of human P2RY13 (AA: 1-49) expresed in E. Coli
Molecular Weight of Antigen 40.8 kDa
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 53829
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.