Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P2Y13/P2RY13/GPR86 Antibody (6G12E10) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP252526
Description
P2Y13/P2RY13/GPR86 Monoclonal specifically detects P2Y13/P2RY13/GPR86 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
P2Y13/P2RY13/GPR86 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
G protein-coupled receptor 86, GPR86GPCR1, GPR94, G-protein coupled receptor 86, G-protein coupled receptor 94, P2Y purinoceptor 13, P2Y13FKSG77, purinergic receptor P2Y, G-protein coupled, 13, SP174 | |
Mouse | |
40.8 kDa | |
0.1 mg | |
GPCR | |
53829 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Flow Cytometry, ELISA, CyTOF | |
6G12E10 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
P2RY13 | |
Purified recombinant fragment of human P2RY13 (AA: 1-49) expresed in E. Coli | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction